missing translation for 'onlineSavingsMsg'
Learn More

4'-Fluoro-2'-methylacetophenone, 97%, Thermo Scientific™

Product Code. 11336959
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11336959 1 g 1g
11346959 5 g 5g
11356959 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11336959 Supplier Thermo Scientific Alfa Aesar Supplier No. H26823.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 446-29-7
Molecular Formula C9H9FO
Molecular Weight (g/mol) 152.17
MDL Number MFCD00028041
InChI Key GCTZPACJEUDZAX-UHFFFAOYSA-N
PubChem CID 3494772
IUPAC Name 1-(4-fluoro-2-methylphenyl)ethanone
SMILES CC(=O)C1=C(C)C=C(F)C=C1

Specifications

Quantity 1 g
Synonym 1-4-fluoro-2-methylphenyl ethanone, 4'-fluoro-2'-methylacetophenone, 1-4-fluoro-2-methylphenyl ethan-1-one, ethanone, 1-4-fluoro-2-methylphenyl, 4'-fluoro-2'-methyacetophenone, 1-acetyl-4-fluoro-2-methylbenzene, ethanone,1-4-fluoro-2-methylphenyl
Formula Weight 152.17
Percent Purity 97%
Chemical Name or Material 4'-Fluoro-2'-methylacetophenone
Product Title
Select an issue

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.