accessibility menu, dialog, popup

UserName

CH3FOCH3

CAS RN 446-29-7

IUPAC Name: 1-(4-fluoro-2-methylphenyl)ethan-1-one
Synonyms: 1-(4-fluoro-2-methylphenyl)ethanone
Molecular Weight (g/mol): 152.17
Molecular Formula: C9H9FO
InChi Key: GCTZPACJEUDZAX-UHFFFAOYSA-N
SMILES: CC(=O)C1=C(C)C=C(F)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results