Learn More
4-Ethynyl-2,6-dimethyl-4-heptanol, 97%, Thermo Scientific™
Product Code. 11323166
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
2 g
10 g
missing translation for 'unitSize':
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity missing translation for 'unitSize'
11323166 2 g 2g
11333166 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Product Code. 11323166 missing translation for 'mfr' Thermo Scientific Alfa Aesar missing translation for 'supplierNo' L12446.04
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 10562-68-2
Molecular Formula C11H20O
Molecular Weight (g/mol) 168.28
MDL Number MFCD00041605
InChI Key CSNWKQHTZXPWJS-UHFFFAOYSA-N
Synonym 4-ethynyl-2,6-dimethyl-4-heptanol, acmc-1bvp6, 2,6-dimethyl-4-ethynyl-4-heptanol, 4-heptanol,4-ethynyl-2,6-dimethyl, 5-methyl-3-2-methylpropyl hex-1-yn-3-ol
PubChem CID 7010451
IUPAC Name 4-ethynyl-2,6-dimethylheptan-4-ol
SMILES CC(C)CC(O)(CC(C)C)C#C
Specifications
Boiling Point 83°C to 84°C (18 mmHg)
Refractive Index 1.442
Quantity 2 g
Beilstein 1752640
Formula Weight 168.28
Percent Purity 97%
Chemical Name or Material 4-Ethynyl-2,6-dimethyl-4-heptanol