missing translation for 'onlineSavingsMsg'
Learn More

4-Ethyl-4-formylhexanenitrile, 95%, Thermo Scientific™

Product Code. p-130063 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10163780 5 g Glass bottle 5g
10666402 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10163780 Supplier Thermo Scientific Acros Supplier No. 195570050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2938-69-4
Molecular Formula C9H15NO
Molecular Weight (g/mol) 153.22
MDL Number MFCD00047654
InChI Key AMUXIQHQXOKMTN-UHFFFAOYSA-N
Synonym 4-cyano-2,2-diethylbutyraldehyde, acmc-1cbzp, 4-cyano-2,2-diethylbutanal, hexanenitrile,4-ethyl-4-formyl, hexanenitrile, 4-ethyl-4-formyl, 3-2-cyanoethyl-3-formylpentane, 2-beta-cyanoethyl-2-ethyl butyraldehyde
PubChem CID 76248
IUPAC Name 4-ethyl-4-formylhexanenitrile
SMILES CCC(CC)(CCC#N)C=O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9480g/mL
Boiling Point 94°C to 99°C (1.0 mmHg)
Flash Point 96°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula (C2H5)2C(CHO)CH2CH2CN
Refractive Index 1.4505 to 1.4525
Quantity 5 g
Specific Gravity 0.948
Formula Weight 153.22
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 4-Ethyl-4-formylhexanenitrile, 95%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.