accessibility menu, dialog, popup

UserName

CH3H3CNO

CAS RN 2938-69-4

IUPAC Name: 4-ethyl-4-formylhexanenitrile
Synonyms: 4-ethyl-4-formylhexanenitrile
Molecular Weight (g/mol): 153.23
Molecular Formula: C9H15NO
InChi Key: AMUXIQHQXOKMTN-UHFFFAOYSA-N
SMILES: CCC(CC)(CCC#N)C=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)