missing translation for 'onlineSavingsMsg'
Learn More

4-(Dimethylamino)thiobenzamide, 97%, Thermo Scientific™

Product Code. 15487347
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15487347 5 g 5g
15477347 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15487347 Supplier Thermo Scientific Alfa Aesar Supplier No. H34408.06
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-(Dimethylamino)thiobenzamide is used as pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Air sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4714-69-6
Molecular Formula C9H12N2S
Molecular Weight (g/mol) 180.269
MDL Number MFCD03426573
InChI Key VBQDGXAMTXINAS-UHFFFAOYSA-N
Synonym 4-dimethylaminothiobenzamide, 4-dimethylamino benzenecarbothioamide, 4-dimethylamino-thiobenzamide, 4-dimethylamino thiobenzamide, 4-dimethylamino benzene-1-carbothioamide, benzenecarbothioamide, 4-dimethylamino, 4-dimethylamino benzothioamide, 4-n,n-dimethylaminothiobenzamide, p-dimethylaminobenzothiamide, p-dimethylaminothiobenzamide
PubChem CID 816768
IUPAC Name 4-(dimethylamino)benzenecarbothioamide
SMILES CN(C)C1=CC=C(C=C1)C(=S)N

Specifications

Melting Point 241°C to 245°C
Quantity 5 g
Sensitivity Air sensitive
Solubility Information Soluble in water.
Formula Weight 180.27
Percent Purity 97%
Chemical Name or Material 4-(Dimethylamino)thiobenzamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.