CAS RN 4714-69-6
IUPAC Name:
4-(dimethylamino)benzene-1-carbothioamide
Synonyms:
4-(dimethylamino)benzenecarbothioamide
Molecular Weight (g/mol):
180.27
Molecular Formula:
C9H12N2S
InChi Key:
VBQDGXAMTXINAS-UHFFFAOYSA-N
SMILES:
CN(C)C1=CC=C(C=C1)C(N)=S
1
–
1
of
1
results
1
4-(Dimethylamino)thiobenzamide, 97%, Thermo Scientific™
CAS: 4714-69-6 Molecular Formula: C9H12N2S Molecular Weight (g/mol): 180.269 MDL Number: MFCD03426573 InChI Key: VBQDGXAMTXINAS-UHFFFAOYSA-N Synonym: 4-dimethylaminothiobenzamide,4-dimethylamino benzenecarbothioamide,4-dimethylamino-thiobenzamide,4-dimethylamino thiobenzamide,4-dimethylamino benzene-1-carbothioamide,benzenecarbothioamide, 4-dimethylamino,4-dimethylamino benzothioamide,4-n,n-dimethylaminothiobenzamide,p-dimethylaminobenzothiamide,p-dimethylaminothiobenzamide PubChem CID: 816768 IUPAC Name: 4-(dimethylamino)benzenecarbothioamide SMILES: CN(C)C1=CC=C(C=C1)C(=S)N