missing translation for 'onlineSavingsMsg'
Learn More

4-Cyano-4'-n-hexyloxybiphenyl, 97%, Thermo Scientific™

Product Code. p-7037208
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15412268 1 g 1g
15422268 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15412268 Supplier Thermo Scientific Alfa Aesar Supplier No. H55876.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Cyano-4′-n-hexyloxybiphenyl, is used as a intermediate of liquid crystals. It is mainly used as the pharmaceutical intermediate, liquid crystal intermediate, liquid crystal monomers.

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41424-11-7
Molecular Formula C19H21NO
Molecular Weight (g/mol) 279.38
MDL Number MFCD00134145
InChI Key YUYXUPYNSOFWFV-UHFFFAOYSA-N
Synonym 4'-hexyloxy-4-biphenylcarbonitrile, 6ocb, liquid crystal 549, 1,1'-biphenyl-4-carbonitrile, 4'-hexyloxy, 4-hexyloxycyanodiphenyl, 4-cyano-4'-hexoxybiphenyl, 4-4-hexyloxyphenyl benzonitrile, 4'-hexyloxy-1,1'-biphenyl-4-carbonitrile
PubChem CID 162465
IUPAC Name 4-(4-hexoxyphenyl)benzonitrile
SMILES CCCCCCOC1=CC=C(C=C1)C1=CC=C(C=C1)C#N

Specifications

Melting Point 56°C to 58°C
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 279.38
Percent Purity 97%
Chemical Name or Material 4-Cyano-4'-n-hexyloxybiphenyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.