accessibility menu, dialog, popup

UserName

H3CON

CAS RN 41424-11-7

IUPAC Name: 4'-(hexyloxy)-[1,1'-biphenyl]-4-carbonitrile
Synonyms: 4'-(hexyloxy)-[1,1'-biphenyl]-4-carbonitrile
Molecular Weight (g/mol): 279.38
Molecular Formula: C19H21NO
InChi Key: YUYXUPYNSOFWFV-UHFFFAOYSA-N
SMILES: CCCCCCOC1=CC=C(C=C1)C1=CC=C(C=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results