missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-fluorotoluene, 99%, Thermo Scientific™

Product Code. 11447806
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
Unit Size:
100g
250g
25g
Product Code. Quantity unitSize
11447806 250 g 250g
11354759 25 g 25g
11364759 100 g 100g
3 options
This item is not returnable. View return policy

Product Code. 11447806

Brand: Thermo Scientific Alfa Aesar A14753.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 452-75-5
Molecular Formula C7H6ClF
Molecular Weight (g/mol) 144.57
MDL Number MFCD00000571
InChI Key MKFCYQTVSDCXAQ-UHFFFAOYSA-N
Synonym 4-chloro-2-fluorotoluene, 2-fluoro-4-chlorotoluene, benzene, 4-chloro-2-fluoro-1-methyl, pubchem3437, acmc-209k1n, ksc493m8j, mkfcyqtvsdcxaq-uhfffaoysa, 4-chloro-2-fluoro-1-methyl-benzene
PubChem CID 520630
IUPAC Name 4-chloro-2-fluoro-1-methylbenzene
SMILES CC1=CC=C(Cl)C=C1F

Specifications

Density 1.184
Boiling Point 158°C to 160°C
Flash Point 51°C (123°F)
Odor Aromatic
Refractive Index 1.498
Quantity 250 g
UN Number UN1993
Beilstein 1931682
Formula Weight 144.58
Percent Purity 99%
Chemical Name or Material 4-Chloro-2-fluorotoluene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.