accessibility menu, dialog, popup

UserName

CAS RN 452-75-5

CH3ClF

CAS RN 452-75-5

IUPAC Name: 4-chloro-2-fluoro-1-methylbenzene
Synonyms: 4-chloro-2-fluoro-1-methylbenzene
Molecular Weight (g/mol): 144.57
Molecular Formula: C7H6ClF
InChi Key: MKFCYQTVSDCXAQ-UHFFFAOYSA-N
SMILES: CC1=CC=C(Cl)C=C1F
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results