Learn More
4'-Bromononanophenone, 98%, Thermo Scientific™
Product Code. 11485650
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. 11485650
Brand: Thermo Scientific Alfa Aesar H25900.14
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 126334-26-7
Molecular Formula C15H21BrO
Molecular Weight (g/mol) 297.236
MDL Number MFCD09953463
InChI Key JIVVSHXGCFTTTG-UHFFFAOYSA-N
Synonym 1-4-bromophenyl nonan-1-one, 4'-bromononanophenone, 1-nonanone,1-4-bromophenyl, acmc-20e7mb, 4-nonanoyl bromobenzene, 4-bromo-alpha-octylbenzaldehyde
PubChem CID 23117403
IUPAC Name 1-(4-bromophenyl)nonan-1-one
SMILES CCCCCCCCC(=O)C1=CC=C(C=C1)Br
Specifications
Melting Point 65°C to 69°C
Boiling Point 138°C to 140°C (0.1 mmHg)
Quantity 25 g
Formula Weight 297.24
Percent Purity 98%
Chemical Name or Material 4'-Bromononanophenone