CAS RN 126334-26-7
CAS RN 126334-26-7
IUPAC Name:
1-(4-bromophenyl)nonan-1-one
Synonyms:
1-(4-bromophenyl)nonan-1-one
Molecular Weight (g/mol):
297.24
Molecular Formula:
C15H21BrO
InChi Key:
JIVVSHXGCFTTTG-UHFFFAOYSA-N
SMILES:
CCCCCCCCC(=O)C1=CC=C(Br)C=C1
1
–
1
of
1
results
1
4'-Bromononanophenone, 98%, Thermo Scientific™
CAS: 126334-26-7 Molecular Formula: C15H21BrO Molecular Weight (g/mol): 297.236 MDL Number: MFCD09953463 InChI Key: JIVVSHXGCFTTTG-UHFFFAOYSA-N Synonym: 1-4-bromophenyl nonan-1-one,4'-bromononanophenone,1-nonanone,1-4-bromophenyl,acmc-20e7mb,4-nonanoyl bromobenzene,4-bromo-alpha-octylbenzaldehyde PubChem CID: 23117403 IUPAC Name: 1-(4-bromophenyl)nonan-1-one SMILES: CCCCCCCCC(=O)C1=CC=C(C=C1)Br