Learn More
4-Bromo-N-(2,4-dimethylphenyl)benzamide, 97%, Thermo Scientific™
Product Code. 15483788
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15483788 1 g 1g
15493788 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15483788 Supplier Thermo Scientific Alfa Aesar Supplier No. H57284.03
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 282091-66-1
Molecular Formula C15H14BrNO
Molecular Weight (g/mol) 304.19
MDL Number MFCD00436496
InChI Key CGUNDMXTDHRITR-UHFFFAOYSA-N
Synonym 4-bromo-n-2,4-dimethylphenyl benzamide, 4-bromo-n-2,4-dimethyl-phenyl-benzamide
PubChem CID 774375
IUPAC Name 4-bromo-N-(2,4-dimethylphenyl)benzamide
SMILES CC1=CC(C)=C(NC(=O)C2=CC=C(Br)C=C2)C=C1
Specifications
Quantity 1 g
Formula Weight 304.19
Percent Purity 97%
Chemical Name or Material 4-Bromo-N-(2,4-dimethylphenyl)benzamide