accessibility menu, dialog, popup

UserName

CH3HNOBrCH3

CAS RN 282091-66-1

IUPAC Name: 4-bromo-N-(2,4-dimethylphenyl)benzamide
Synonyms: 4-bromo-N-(2,4-dimethylphenyl)benzamide
Molecular Weight (g/mol): 304.19
Molecular Formula: C15H14BrNO
InChi Key: CGUNDMXTDHRITR-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(NC(=O)C2=CC=C(Br)C=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results