Learn More
4-Bromo-3-chloroaniline, 96%
Product Code. 11312086
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11312086 50 g 50g
11302086 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11312086 Supplier Thermo Scientific Alfa Aesar Supplier No. L03268.18
Request Bulk or Custom Format
CAS: 21402-26-6 | C6H5BrClN | 206.467 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 21402-26-6
Molecular Formula C6H5BrClN
Molecular Weight (g/mol) 206.467
MDL Number MFCD00041311
InChI Key QLYHPNUFNZJXOQ-UHFFFAOYSA-N
Synonym benzenamine, 4-bromo-3-chloro, 3-chloro-4-bromoaniline, unii-2gtm4sw4oh, 2gtm4sw4oh, 4-bromo-3-chloro-aniline, 4-bromo-3-chlorophenyl amine, pubchem3579, zlchem 1051, 4-bromo-3-chloro aniline, 4-bromo-3-chlorophenylamine
PubChem CID 88888
IUPAC Name 4-bromo-3-chloroaniline
SMILES C1=CC(=C(C=C1N)Cl)Br
Specifications
Melting Point 61°C to 66°C
Odor Characteristic
Quantity 50 g
UN Number UN2811
Beilstein 1210160
Formula Weight 206.47
Percent Purity 96%
Chemical Name or Material 4-Bromo-3-chloroaniline