accessibility menu, dialog, popup

UserName

NH2BrCl

CAS RN 21402-26-6

IUPAC Name: 4-bromo-3-chloroaniline
Synonyms: 4-bromo-3-chloroaniline
Molecular Weight (g/mol): 206.47
Molecular Formula: C6H5BrClN
InChi Key: QLYHPNUFNZJXOQ-UHFFFAOYSA-N
SMILES: NC1=CC=C(Br)C(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results