Learn More
4-Bromo-3,5-dimethyl-1-(2-tetrahydropyranyl)-1H-pyrazole, 95%, Thermo Scientific™
Product Code. 15438255
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. 15438255
Brand: Thermo Scientific Alfa Aesar H32968.MD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1187385-69-8
Molecular Formula C10H15BrN2O
Molecular Weight (g/mol) 259.147
MDL Number MFCD11844792
InChI Key SQNKVHBFAOKTBH-UHFFFAOYSA-N
Synonym 4-bromo-3,5-dimethyl-1-tetrahydropyran-2-yl pyrazole, 4-bromo-3,5-dimethyl-1-tetrahydro-2h-pyran-2-yl-1h-pyrazole, 4-bromo-3,5-dimethyl-1-oxan-2-yl-1h-pyrazole, 4-bromo-3,5-dimethyl-1-2-tetrahydropyranyl-1h-pyrazole, 4-bromo-3,5-dimethyl-1-oxan-2-yl pyrazole, acmc-2099yc
PubChem CID 45588171
IUPAC Name 4-bromo-3,5-dimethyl-1-(oxan-2-yl)pyrazole
SMILES CC1=C(C(=NN1C2CCCCO2)C)Br
Specifications
Quantity 250 mg
Formula Weight 259.15
Percent Purity 95%
Chemical Name or Material 4-Bromo-3,5-dimethyl-1-(2-tetrahydropyranyl)-1H-pyrazole