accessibility menu, dialog, popup

UserName

CAS RN 1187385-69-8

CH3NNOCH3Br

CAS RN 1187385-69-8

IUPAC Name: 4-bromo-3,5-dimethyl-1-(oxan-2-yl)-1H-pyrazole
Synonyms: 4-bromo-3,5-dimethyl-1-(oxan-2-yl)pyrazole
Molecular Weight (g/mol): 259.15
Molecular Formula: C10H15BrN2O
InChi Key: SQNKVHBFAOKTBH-UHFFFAOYNA-N
SMILES: CC1=NN(C2CCCCO2)C(C)=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results