Learn More
4-Amino-5-bromo-6-chloropyrimidine, 98%, Thermo Scientific™
Product Code. 15431539
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Product Code. Quantity unitSize
15431539 250 mg 250mg
15411539 1 g 1g
15421539 5 g 5g
3 options
Product Code. 15431539
Brand: Thermo Scientific Alfa Aesar H64369.MD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 663193-80-4
Molecular Formula C4H3BrClN3
Molecular Weight (g/mol) 208.443
MDL Number MFCD09999224
InChI Key DQSBRYJETNYGCI-UHFFFAOYSA-N
Synonym 4-amino-5-bromo-6-chloropyrimidine, 5-bromo-6-chloro-4-pyrimidinamine, 5-bromo-6-chloro-pyrimidin-4-ylamine
PubChem CID 45789713
IUPAC Name 5-bromo-6-chloropyrimidin-4-amine
SMILES C1=NC(=C(C(=N1)Cl)Br)N
Specifications
Color Gray
Quantity 250 mg
Beilstein 23483365
Formula Weight 208.44
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 4-Amino-5-bromo-6-chloropyrimidine