accessibility menu, dialog, popup

UserName

NNClBrH2N

CAS RN 663193-80-4

IUPAC Name: 5-bromo-6-chloropyrimidin-4-amine
Synonyms: 5-bromo-6-chloropyrimidin-4-amine
Molecular Weight (g/mol): 208.44
Molecular Formula: C4H3BrClN3
InChi Key: DQSBRYJETNYGCI-UHFFFAOYSA-N
SMILES: NC1=C(Br)C(Cl)=NC=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results