Learn More
Memmert™ 4 - 20 mA Current Loop Interface
Product Code. 17119974 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
ICO105
ICO150
ICO240
ICO50
Unit Size:
Each
Product Code. 17119974
Brand: Memmert™ ICO150V3
Only null left
Add to basket
Add to basket
4-20mA current loop interface Temperature controller, actual value
  • Temperature controller
  • Actual value; 0 to +70°C = 4 - 20 mA
Specifications
For Use With (Equipment) ICO150
Type Temperature Controller