Learn More
Memmert™ 4 - 20 mA Current Loop Interface
Product Code. p-7220899 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
HPP1060eco
HPP110eco
HPP1400eco
HPP2200eco
HPP260eco
HPP410eco
HPP750eco
IPP1060eco
IPP110eco
IPP1400eco
IPP2200eco
IPP260eco
IPP410eco
IPP750eco
Unit Size:
Each
Product Code. 17109734
Brand: Memmert™ HPP1400ecoV3
Only null left
Add to basket
Add to basket
4-20mA current loop interface temperature controller
  • Temperature controller
  • Actual value; -10 to +80 °C = 4 -20 mA
Specifications
For Use With (Equipment) HPP1400eco
Type Temperature Controller