Learn More
Memmert™ 4 - 20 mA Current Loop Interface
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
HPP1060eco
HPP110eco
HPP1400eco
HPP2200eco
HPP260eco
HPP410eco
HPP750eco
IPP1060eco
IPP110eco
IPP1400eco
IPP2200eco
IPP260eco
IPP410eco
IPP750eco
Unit Size:
Each
Description
- Temperature controller
- Actual value; -10 to +80 °C = 4 -20 mA
Specifications
Specifications
| For Use With (Equipment) | IPP750eco |
| Type | Temperature Controller |