Learn More
Memmert™ 4 - 20 mA Current Loop Interface
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
ICO105
ICO150
ICO240
ICO50
Unit Size:
Each
Description
- Requires option K7 or K6
- Humidity controller
- Actual value (0 to 100 % rh = 4 - 20 mA)
Specifications
Specifications
| Description | Requires option K7 or K6 |
| For Use With (Equipment) | ICO105 |