Learn More
4-(2-Benzothiazolyl)aniline, 98%
Product Code. 15455837
Change view
Click to view available options
Quantity:
1 g
5 g
Packungsgröße:
1g
5g
Artikelnummer. Quantity unitSize
15455837 1 g 1g
15465837 5 g 5g
2 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 15455837
Marke: Thermo Scientific Alfa Aesar H33052.03
Request Bulk or Custom Format
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
CAS: 6278-73-5 | C13H10N2S | 226.30 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemische Identifikatoren
CAS 6278-73-5
Molecular Formula C13H10N2S
Molecular Weight (g/mol) 226.30
MDL Number MFCD00182896
InChI Key WKRCOZSCENDENK-UHFFFAOYSA-N
Synonym 4-1,3-benzothiazol-2-yl aniline, 4-benzo d thiazol-2-yl aniline, 4-benzothiazol-2-yl-phenylamine, 2-4-aminophenyl benzothiazole, 4-1,3-benzothiazol-2-yl phenylamine, 4-benzothiazol-2-ylphenylamine, benzenamine, 4-2-benzothiazolyl, chembl93118, 4-1,3-benzothiazol-2-yl phenyl amine, benzenamine,4-2-benzothiazolyl
PubChem CID 234475
IUPAC Name 4-(1,3-benzothiazol-2-yl)aniline
SMILES NC1=CC=C(C=C1)C1=NC2=CC=CC=C2S1
Spezifikation
Melting Point 152°C to 154°C
Quantity 1 g
Formula Weight 226.3
Percent Purity 98%
Chemical Name or Material 4-(2-Benzothiazolyl)aniline