CAS RN 6278-73-5
CAS RN 6278-73-5
IUPAC Name:
4-(1,3-benzothiazol-2-yl)aniline
Synonyms:
4-(1,3-benzothiazol-2-yl)aniline
Molecular Weight (g/mol):
226.3
Molecular Formula:
C13H10N2S
InChi Key:
WKRCOZSCENDENK-UHFFFAOYSA-N
SMILES:
NC1=CC=C(C=C1)C1=NC2=CC=CC=C2S1
1
–
1
of
1
results
1
4-(2-Benzothiazolyl)aniline, 98%
CAS: 6278-73-5 Molecular Formula: C13H10N2S Molecular Weight (g/mol): 226.30 MDL Number: MFCD00182896 InChI Key: WKRCOZSCENDENK-UHFFFAOYSA-N Synonym: 4-1,3-benzothiazol-2-yl aniline,4-benzo d thiazol-2-yl aniline,4-benzothiazol-2-yl-phenylamine,2-4-aminophenyl benzothiazole,4-1,3-benzothiazol-2-yl phenylamine,4-benzothiazol-2-ylphenylamine,benzenamine, 4-2-benzothiazolyl,chembl93118,4-1,3-benzothiazol-2-yl phenyl amine,benzenamine,4-2-benzothiazolyl PubChem CID: 234475 IUPAC Name: 4-(1,3-benzothiazol-2-yl)aniline SMILES: NC1=CC=C(C=C1)C1=NC2=CC=CC=C2S1