Learn More
RETSCH 360° Stainless Steel Ring Sieve For SR200/SR300 Rotor Beater Mills
Product Code. 10557791 Shop All Retsch Products
Change view
Click to view available options
Pore Size:
80 μm
1 mm
120 μm
1.25 mm
1.5 mm
2 mm
200 μm
250 μm
3 mm
4 mm
5 mm
500 μm
6 mm
750 μm
8 mm
10 mm
Unit Size:
Each
Product Code. Pore Size unitSize
10557791 500 μm Each
10778781 80 μm Each
10517221 120 μm Each
10242401 250 μm Each
10191161 200 μm Each
10394191 750 μm Each
10262401 1.25 mm Each
10597981 1.5 mm Each
10354481 10 mm Each
10354241 1 mm Each
10314341 3 mm Each
10161261 2 mm Each
10420263 4 mm Each
10769551 5 mm Each
10121361 6 mm Each
10212501 8 mm Each
16 options
Product Code. 10557791
Brand: RETSCH 02.407.0030
Only null left
Specifications
Quantity 1Each
Pore Size 500 μm
Type Ring Sieve
Material Stainless Steel
For Use With (Equipment) SR200/SR300 Rotor Beater Mills