Learn More
- Home
- Products
- Filters and Filtration
- Mechanical Sieves
- RETSCH 305 dia. x 40 mmH Stainless Steel Test Sieve with Square Holes
RETSCH 305 dia. x 40 mmH Stainless Steel Test Sieve with Square Holes
Product Code. 10378272 Shop All Retsch Products
Change view
Click to view available options
Pore Size:
4 mm
4.50 mm
4.75 mm
5 mm
5.60 mm
6.30 mm
6.70 mm
7.10 mm
8 mm
9 mm
9.50 mm
10 mm
11.2 mm
12.5 mm
13.2 mm
14 mm
16 mm
18 mm
19 mm
20 mm
22.4 mm
25 mm
26.5 mm
28 mm
31.5 mm
35.5 mm
37.5 mm
40 mm
45 mm
50 mm
53 mm
56 mm
63 mm
71 mm
75 mm
80 mm
90 mm
100 mm
106 mm
112 mm
125 mm
Unit Size:
Each
| Product Code. | Pore Size | unitSize |
|---|---|---|
| 10378272 | 106 mm | Each |
| 10614403 | 100 mm | Each |
| 10174922 | 10 mm | Each |
| 10719964 | 11.2 mm | Each |
| 10555173 | 12.5 mm | Each |
| 10614593 | 112 mm | Each |
| 10114542 | 125 mm | Each |
| 10543633 | 18 mm | Each |
| 10173812 | 20 mm | Each |
| 10482652 | 13.2 mm | Each |
| 10684583 | 19 mm | Each |
| 10226842 | 16 mm | Each |
| 10740125 | 14 mm | Each |
| 10368462 | 22.4 mm | Each |
| 10799924 | 25 mm | Each |
| 10123962 | 26.5 mm | Each |
| 10584203 | 28 mm | Each |
| 10327892 | 31.5 mm | Each |
| 10092982 | 4.50 mm | Each |
| 10524593 | 37.5 mm | Each |
| 10022832 | 35.5 mm | Each |
| 10788794 | 4.75 mm | Each |
| 10337892 | 40 mm | Each |
| 10616703 | 45 mm | Each |
| 10695933 | 5.60 mm | Each |
| 10002682 | 50 mm | Each |
| 10574973 | 53 mm | Each |
| 10082982 | 4 mm | Each |
| 10266552 | 56 mm | Each |
| 10444572 | 5 mm | Each |
| 10002862 | 6.30 mm | Each |
| 10062842 | 6.70 mm | Each |
| 10664013 | 63 mm | Each |
| 10534023 | 7.10 mm | Each |
| 10328132 | 71 mm | Each |
| 10072722 | 75 mm | Each |
| 10505173 | 80 mm | Each |
| 10720125 | 8 mm | Each |
| 10646323 | 9.50 mm | Each |
| 10266172 | 90 mm | Each |
| 10594593 | 9 mm | Each |
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Specifications
Specifications
| Quantity | 1Each |
| Pore Size | 106 mm |
| Type | Test Sieve |
| Certifications/Compliance | ISO 3310/2 |
| Material | Stainless Steel |
| Height (Metric) | 40mm |
| For Use With (Application) | For separation, fractioning, particle size determination |
| Diameter (Metric) Outer | 305 mm |
| Frame Material | Stainless Steel |
| Mesh Material | Stainless Steel |