Learn More
RETSCH 305 dia. x 40 mmH Stainless Steel Test Sieve with Square Holes
Product Code. 10266172 Shop All Retsch Products
Change view
Click to view available options
Pore Size:
4 mm
4.50 mm
4.75 mm
5 mm
5.60 mm
6.30 mm
6.70 mm
7.10 mm
8 mm
9 mm
9.50 mm
10 mm
11.2 mm
12.5 mm
13.2 mm
14 mm
16 mm
18 mm
19 mm
20 mm
22.4 mm
25 mm
26.5 mm
28 mm
31.5 mm
35.5 mm
37.5 mm
40 mm
45 mm
50 mm
53 mm
56 mm
63 mm
71 mm
75 mm
80 mm
90 mm
100 mm
106 mm
112 mm
125 mm
Unit Size:
Each
Product Code. Pore Size unitSize
10266172 90 mm Each
10614403 100 mm Each
10378272 106 mm Each
10174922 10 mm Each
10719964 11.2 mm Each
10555173 12.5 mm Each
10614593 112 mm Each
10114542 125 mm Each
10543633 18 mm Each
10173812 20 mm Each
10482652 13.2 mm Each
10684583 19 mm Each
10226842 16 mm Each
10740125 14 mm Each
10368462 22.4 mm Each
10799924 25 mm Each
10123962 26.5 mm Each
10584203 28 mm Each
10327892 31.5 mm Each
10092982 4.50 mm Each
10524593 37.5 mm Each
10022832 35.5 mm Each
10788794 4.75 mm Each
10337892 40 mm Each
10616703 45 mm Each
10695933 5.60 mm Each
10002682 50 mm Each
10574973 53 mm Each
10082982 4 mm Each
10266552 56 mm Each
10444572 5 mm Each
10002862 6.30 mm Each
10062842 6.70 mm Each
10664013 63 mm Each
10534023 7.10 mm Each
10328132 71 mm Each
10072722 75 mm Each
10505173 80 mm Each
10720125 8 mm Each
10646323 9.50 mm Each
10594593 9 mm Each
41 options
Product Code. 10266172
Brand: RETSCH 60.138.090000
Only null left
Specifications
Quantity 1Each
Pore Size 90 mm
Type Test Sieve
Certifications/Compliance ISO 3310/2
Material Stainless Steel
Height (Metric) 40mm
For Use With (Application) For separation, fractioning, particle size determination
Diameter (Metric) Outer 305 mm
Frame Material Stainless Steel
Mesh Material Stainless Steel