Learn More
RETSCH 305 dia. x 40 mmH Stainless Steel Test Sieve, ISO Certified, Pore Sizes: Under 10 mm
Product Code. p-8003368 Shop All Retsch Products
Change view
Click to view available options
Pore Size:
1 mm
1.12 mm
1.18 mm
1.25 mm
1.40 mm
1.60 mm
1.70 mm
1.80 mm
2 mm
2.24 mm
2.36 mm
2.50 mm
2.80 mm
3.15 mm
3.35 mm
3.55 mm
4 mm
4.50 mm
4.75 mm
5 mm
5.60 mm
6.30 mm
6.70 mm
7.10 mm
8 mm
9 mm
9.50 mm
Unit Size:
Each
Product Code. Pore Size unitSize
10085081 1.12 mm Each
10711243 1.18 mm Each
10711433 1.40 mm Each
10584872 1.25 mm Each
10219171 1.60 mm Each
10487961 1.80 mm Each
10074931 1.70 mm Each
10310822 1 mm Each
10034971 2.24 mm Each
10739322 2.36 mm Each
10370382 2.50 mm Each
10218501 2.80 mm Each
10116461 2 mm Each
10710093 3.15 mm Each
10478731 3.35 mm Each
10478921 3.55 mm Each
10156881 4.50 mm Each
10593722 4.75 mm Each
10166881 5.60 mm Each
10720093 4 mm Each
10075051 5 mm Each
10054911 6.30 mm Each
10157121 7.10 mm Each
10630642 9 mm Each
10004951 9.50 mm Each
10064911 8 mm Each
10486621 6.70 mm Each
27 options
Product Code. 10085081
Brand: RETSCH 60.158.001120
Only null left
Specifications
Quantity 1Each
Pore Size 1.12 mm
Type Test Sieve
Certifications/Compliance ISO 3310/1
Material Stainless Steel
For Use With (Equipment) Shaker AS 300
Includes Certificate indicating Compliance
Height (Metric) 40mm
For Use With (Application) For separation, fractioning, particle size determination
Diameter (Metric) Outer 305 mm
Frame Material Stainless Steel
Mesh Material Stainless Steel