Learn More
RETSCH 305 dia. x 40 mmH Stainless Steel Test Sieve, ASTM Certified, Micrometer Pore Sizes
Product Code. 10248831 Shop All Retsch Products
Change view
Click to view available options
Pore Size:
25 μm
32 μm
38 μm
45 μm
53 μm
63 μm
75 μm
106 μm
125 μm
150 μm
212 μm
250 μm
300 μm
355 μm
425 μm
500 μm
600 μm
710 μm
850 μm
Unit Size:
Each
Product Code. Pore Size unitSize
10248831 38 μm Each
10095081 125 μm Each
10310962 106 μm Each
10005091 150 μm Each
10525262 212 μm Each
10279211 250 μm Each
10085011 25 μm Each
10268821 355 μm Each
10085021 32 μm Each
10535262 300 μm Each
10441952 500 μm Each
10553522 425 μm Each
10258831 45 μm Each
10156171 53 μm Each
10449841 600 μm Each
10035051 63 μm Each
10534302 75 μm Each
10196061 850 μm Each
10024761 710 μm Each
19 options
Product Code. 10248831
Brand: RETSCH 60.159.000038
Only null left
Specifications
Quantity 1Each
Pore Size 38 μm
Type Test Sieve
Certifications/Compliance ASTM E 11
Material Stainless Steel
For Use With (Equipment) Shaker AS 300
Height (Metric) 40mm
For Use With (Application) For separation, fractioning, particle size determination
Diameter (Metric) Outer 305 mm
Frame Material Stainless Steel
Mesh Material Stainless Steel