Learn More
- Home
- Products
- Filters and Filtration
- Mechanical Sieves
- RETSCH 305 dia. x 40 mmH Stainless Steel Sieve with Round Holes, Pore Size: 10 mm and Larger
RETSCH 305 dia. x 40 mmH Stainless Steel Sieve with Round Holes, Pore Size: 10 mm and Larger
Product Code. 10432272 Shop All Retsch Products
Change view
Click to view available options
Pore Size:
10 mm
11.2 mm
12.5 mm
13.2 mm
14 mm
16 mm
18 mm
19 mm
20 mm
22.4 mm
25 mm
26.5 mm
28 mm
31.5 mm
35.5 mm
37.5 mm
40 mm
45 mm
50 mm
53 mm
56 mm
63 mm
71 mm
75 mm
80 mm
90 mm
100 mm
106 mm
112 mm
125 mm
Unit Size:
Each
| Product Code. | Pore Size | unitSize |
|---|---|---|
| 10432272 | 19 mm | Each |
| 10368032 | 100 mm | Each |
| 10276552 | 106 mm | Each |
| 10318522 | 10 mm | Each |
| 10544023 | 11.2 mm | Each |
| 10505363 | 112 mm | Each |
| 10544213 | 12.5 mm | Each |
| 10276742 | 125 mm | Each |
| 10144782 | 13.2 mm | Each |
| 10184922 | 16 mm | Each |
| 10646513 | 14 mm | Each |
| 10504603 | 18 mm | Each |
| 10082912 | 20 mm | Each |
| 10482842 | 22.4 mm | Each |
| 10115072 | 25 mm | Each |
| 10125072 | 26.5 mm | Each |
| 10398362 | 28 mm | Each |
| 10729754 | 31.5 mm | Each |
| 10543823 | 35.5 mm | Each |
| 10739754 | 37.5 mm | Each |
| 10378462 | 40 mm | Each |
| 10644973 | 45 mm | Each |
| 10798614 | 56 mm | Each |
| 10645163 | 53 mm | Each |
| 10286262 | 50 mm | Each |
| 10432842 | 63 mm | Each |
| 10708624 | 71 mm | Each |
| 10625743 | 75 mm | Each |
| 10307992 | 80 mm | Each |
| 10473222 | 90 mm | Each |
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Specifications
Specifications
| Quantity | 1Each |
| Pore Size | 19 mm |
| Type | Test Sieve |
| Certifications/Compliance | ISO 3310/2 |
| Material | Stainless Steel |
| Height (Metric) | 40mm |
| For Use With (Application) | For separation, fractioning, particle size determination |
| Diameter (Metric) Outer | 305 mm |
| Frame Material | Stainless Steel |
| Mesh Material | Stainless Steel |