Learn More
Eppendorf™ Research 3 Neo Single Channel Pipettes Ejector Sleeve
Product Code. 30689612 Shop All Eppendorf Products
Click to view available options
Volume (Metric):
2.5 μL
10 μL
20 μL
100 μL
200 μL
300 μL
1000 μL
2 mL
5 mL
10 mL
Unit Size:
Each
Product Code. 30689612
Brand: Eppendorf™ 3102630000
Ejector sleeves allow for easy and efficient tip ejection
  • Ejector sleeves allow for easy and efficient tip ejection
  • These original Eppendorf spare parts are designed to replace the ejector sleeves of Eppendorf Research™ 3 neo pipettes
  • Available in various color code with ejection carrier
TRUSTED_SUSTAINABILITY
Specifications
For Use With Pipettes
Material Polypropylene
Type Ejector Sleeve
Volume (Metric) 2.5 μL
For Use With (Equipment) Pipette Tips
Quantity 1 Pk.