Learn More
Eppendorf™ Research 3 Neo Single Channel Pipettes Ejector Sleeve
Product Code. 30689602 Shop All Eppendorf Products
Change view
Click to view available options
Volume (Metric):
2.5 μL
10 μL
20 μL
100 μL
200 μL
300 μL
1000 μL
2 mL
5 mL
10 mL
Unit Size:
Each
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
30689602 10 μL Each
30689612 2.5 μL Each
30689642 20 μL Each
30689641 100 μL Each
30689679 200 μL Each
30689739 300 μL Each
30689697 1000 μL Each
30689618 2 mL Each
30689728 5 mL Each
30689682 10 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 30689602 Supplier Eppendorf™ Supplier No. 3102630001
Powering Proteomics
Use promo code 28003
View all promotions
In Stock
Add to basket
Add to basket
Ejector sleeves allow for easy and efficient tip ejection
  • Ejector sleeves allow for easy and efficient tip ejection
  • These original Eppendorf spare parts are designed to replace the ejector sleeves of Eppendorf Research™ 3 neo pipettes
  • Available in various color code with ejection carrier
TRUSTED_SUSTAINABILITY
Specifications
For Use With Pipettes
Material Polypropylene
Type Ejector Sleeve
Volume (Metric) 10 μL
For Use With (Equipment) Pipette Tips
Quantity 1 Pk.