Learn More
Eppendorf™ Research 3 Neo Adjustment Tool
Shop All Eppendorf Products
Click to view available options
Type:
Pipette Adjustment Tool
Unit Size:
Pack of 5
Description
- Adjusts factory or temporary settings on Eppendorf Research™ plus and 3 neo pipettes
- Essential for precise calibration and performance optimization
- Original Eppendorf spare part ensures compatibility and reliability
- Ideal replacement if the tool from original packaging is misplaced
- Includes 3 Neo Adjustment Tools per set
Specifications
Specifications
| For Use With | Pipettes |
| Material | Polypropylene |
| Type | Pipette Adjustment Tool |
| For Use With (Equipment) | 3 Neo |
| Quantity | 1 Pk. |