missing translation for 'onlineSavingsMsg'
Learn More

3-Methylcinnamic acid, predominantly trans, 98%, Thermo Scientific™

Product Code. p-7025444
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
This item is not returnable. View return policy

Product Code. 11375339

Brand: Thermo Scientific Alfa Aesar B20914.04

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3029-79-6
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.188
MDL Number MFCD00016844
InChI Key JZINNAKNHHQBOS-AATRIKPKSA-N
Synonym 3-methylcinnamic acid, m-methylcinnamic acid, e-3-m-tolyl acrylic acid, 3-m-tolyl acrylic acid, 2e-3-3-methylphenyl acrylic acid, 2e-3-3-methylphenyl prop-2-enoic acid, cinnamic acid, m-methyl, 3-methylcinnamic acid, predominantly trans, 2e-3-3-methylphenyl-2-propenoic acid, 2-propenoic acid, 3-3-methylphenyl-, 2e
PubChem CID 2063448
ChEBI CHEBI:61118
IUPAC Name (E)-3-(3-methylphenyl)prop-2-enoic acid
SMILES CC1=CC=CC(=C1)C=CC(=O)O

Specifications

Melting Point 115°C to 119°C
Quantity 2 g
Beilstein 2042550
Formula Weight 162.19
Percent Purity 98%
Chemical Name or Material 3-Methylcinnamic acid, predominantly trans
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.