CAS RN 3029-79-6
CAS RN 3029-79-6
IUPAC Name:
(2E)-3-(3-methylphenyl)prop-2-enoate
Synonyms:
(2E)-3-(3-methylphenyl)prop-2-enoate
Molecular Weight (g/mol):
161.18
Molecular Formula:
C10H9O2
InChi Key:
JZINNAKNHHQBOS-AATRIKPKSA-M
SMILES:
CC1=CC=CC(\C=C\C([O-])=O)=C1
1
–
2
of
2
results
1
3-Methylcinnamic acid, predominantly trans, 98%, Thermo Scientific™
CAS: 3029-79-6 Molecular Formula: C10H10O2 Molecular Weight (g/mol): 162.188 MDL Number: MFCD00016844 InChI Key: JZINNAKNHHQBOS-AATRIKPKSA-N Synonym: 3-methylcinnamic acid,m-methylcinnamic acid,e-3-m-tolyl acrylic acid,3-m-tolyl acrylic acid,2e-3-3-methylphenyl acrylic acid,2e-3-3-methylphenyl prop-2-enoic acid,cinnamic acid, m-methyl,3-methylcinnamic acid, predominantly trans,2e-3-3-methylphenyl-2-propenoic acid,2-propenoic acid, 3-3-methylphenyl-, 2e PubChem CID: 2063448 ChEBI: CHEBI:61118 IUPAC Name: (E)-3-(3-methylphenyl)prop-2-enoic acid SMILES: CC1=CC=CC(=C1)C=CC(=O)O