Learn More
3-Methyl-5-(trifluoromethyl)benzyl alcohol, 97%, Thermo Scientific™
Product Code. 15428565
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. 15428565
Brand: Thermo Scientific Alfa Aesar H33655.MD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 116070-38-3
Molecular Formula C9H9F3O
Molecular Weight (g/mol) 190.17
MDL Number MFCD01631605
InChI Key ZGZONWVUWCPMJZ-UHFFFAOYSA-N
Synonym 3-methyl-5-trifluoromethyl benzyl alcohol, 3-methyl-5-trifluoromethyl phenyl methanol, acmc-2099rt, 3-trifluoromethyl-5-methylbenzyl alcohol, 3-methyl-5-trifluoromethyl benzenemethanol, 5-methyl-3-trifluoromethyl phenyl methan-1-ol
PubChem CID 2775612
IUPAC Name [3-methyl-5-(trifluoromethyl)phenyl]methanol
SMILES CC1=CC(CO)=CC(=C1)C(F)(F)F
Specifications
Refractive Index 1.465
Quantity 250 mg
Formula Weight 190.17
Percent Purity 97%
Chemical Name or Material 3-Methyl-5-(trifluoromethyl)benzyl alcohol