CAS RN 116070-38-3
CAS RN 116070-38-3
IUPAC Name:
[3-methyl-5-(trifluoromethyl)phenyl]methanol
Synonyms:
[3-methyl-5-(trifluoromethyl)phenyl]methanol
Molecular Weight (g/mol):
190.17
Molecular Formula:
C9H9F3O
InChi Key:
ZGZONWVUWCPMJZ-UHFFFAOYSA-N
SMILES:
CC1=CC(CO)=CC(=C1)C(F)(F)F
1
–
1
of
1
results
1
3-Methyl-5-(trifluoromethyl)benzyl alcohol, 97%, Thermo Scientific™
CAS: 116070-38-3 Molecular Formula: C9H9F3O Molecular Weight (g/mol): 190.17 MDL Number: MFCD01631605 InChI Key: ZGZONWVUWCPMJZ-UHFFFAOYSA-N Synonym: 3-methyl-5-trifluoromethyl benzyl alcohol,3-methyl-5-trifluoromethyl phenyl methanol,acmc-2099rt,3-trifluoromethyl-5-methylbenzyl alcohol,3-methyl-5-trifluoromethyl benzenemethanol,5-methyl-3-trifluoromethyl phenyl methan-1-ol PubChem CID: 2775612 IUPAC Name: [3-methyl-5-(trifluoromethyl)phenyl]methanol SMILES: CC1=CC(CO)=CC(=C1)C(F)(F)F