Learn More
3-Methyl-2,4-pentanedione, mixture of tautomers, 95%
Product Code. 11324474
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11324474 5 g 5g
11334474 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11324474 Supplier Thermo Scientific Alfa Aesar Supplier No. L01135.06
Request Bulk or Custom Format
CAS: 815-57-6 | C6H10O2 | 114.144 g/mol
3-Methyl-2,4-pentanedione involves in asymmetric substitution of 1,3-diphenyl-2-propenyl acetate catalyzed by amphiphilic resin-supported monodentate phosphine ligands can occur.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Methyl-2,4-pentanedione involves in asymmetric substitution of 1,3-diphenyl-2-propenyl acetate catalyzed by amphiphilic resin-supported monodentate phosphine ligands can occur.
Solubility
Soluble in water.
Notes
Air & heat sensitive. Do not store together with strong bases and oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 815-57-6
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.144
MDL Number MFCD00008762
InChI Key GSOHKPVFCOWKPU-UHFFFAOYSA-N
Synonym 3-methyl-2,4-pentanedione, methylacetylacetone, 1,1-diacetylethane, 2,4-pentanedione, 3-methyl, methyl acetylacetone, 3-acetylbutan-2-one, 3-methyl-acetylacetone, acmc-209plp, 3-methyl-2,4-pentandione, 3-methylpentan-2,4-dione
PubChem CID 69949
IUPAC Name 3-methylpentane-2,4-dione
SMILES CC(C(=O)C)C(=O)C
Specifications
Density 0.98
Boiling Point 170°C to 171°C
Flash Point 56°C (132°F)
Refractive Index 1.442
Quantity 5 g
UN Number UN1224
Beilstein 635909
Solubility Information Soluble in water.
Formula Weight 114.14
Percent Purity 95%
Chemical Name or Material 3-Methyl-2,4-pentanedione, mixture of tautomers