CAS RN 815-57-6
IUPAC Name:
3-methylpentane-2,4-dione
Synonyms:
3-methylpentane-2,4-dione
Molecular Weight (g/mol):
114.14
Molecular Formula:
C6H10O2
InChi Key:
GSOHKPVFCOWKPU-UHFFFAOYSA-N
SMILES:
CC(C(C)=O)C(C)=O
1
–
1
of
1
results
1
3-Methyl-2,4-pentanedione, mixture of tautomers, 95%
CAS: 815-57-6 Molecular Formula: C6H10O2 Molecular Weight (g/mol): 114.144 MDL Number: MFCD00008762 InChI Key: GSOHKPVFCOWKPU-UHFFFAOYSA-N Synonym: 3-methyl-2,4-pentanedione,methylacetylacetone,1,1-diacetylethane,2,4-pentanedione, 3-methyl,methyl acetylacetone,3-acetylbutan-2-one,3-methyl-acetylacetone,acmc-209plp,3-methyl-2,4-pentandione,3-methylpentan-2,4-dione PubChem CID: 69949 IUPAC Name: 3-methylpentane-2,4-dione SMILES: CC(C(=O)C)C(=O)C