Learn More
3-Methoxy-2-methylpyridine, 97%, Thermo Scientific™
Product Code. p-7077431 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 15296038
Brand: Thermo Scientific Acros 453720010
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Chemical Identifiers
CAS 26395-26-6
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.16
InChI Key QRCUQLANPHYVEH-UHFFFAOYSA-N
Synonym 3-methoxy-2-picoline, 3-methoxy-2-methyl-pyridine, pyridine,3-methoxy-2-methyl, pubchem6637, 2-methyl-3-methoxypyridine
PubChem CID 3015270
IUPAC Name 3-methoxy-2-methylpyridine
SMILES CC1=C(C=CC=N1)OC
Specifications
Boiling Point 170.0°C to 175.0°C
Flash Point 61°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 123.16
Percent Purity 97%
Chemical Name or Material 3-Methoxy-2-methylpyridine
compliance-icons
Product Identifier
  • 3-Methoxy-2-methylpyridine, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.