accessibility menu, dialog, popup

UserName

H3CONH3C

CAS RN 26395-26-6

IUPAC Name: 3-methoxy-2-methylpyridine
Synonyms: 3-methoxy-2-methylpyridine
Molecular Weight (g/mol): 123.16
Molecular Formula: C7H9NO
InChi Key: QRCUQLANPHYVEH-UHFFFAOYSA-N
SMILES: COC1=CC=CN=C1C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results