Learn More
(3-Hydroxytricyclo[3.3.1.13,7]dec-1-yl)carbamic Acid 1,1-Dimethylethyl Ester, TRC
Product Code. 30401595
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Product Code. Quantity
30401595 250 mg
30388930 100 mg
30389540 500 mg
3 options
Product Code. 30401595
Brand: Toronto Research Chemicals TRCH971370250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 847795-89-5
Molecular Formula C15H25NO3
Molecular Weight (g/mol) 267.36
Synonym 1,1-Dimethylethyl N-(3-hydroxytricyclo[3.3.1.13,7]dec-1-yl)carbamate (ACI), Carbamic acid, (3-hydroxytricyclo[3.3.1.13,7]dec-1-yl)-, 1,1-dimethylethyl ester (9CI), Carbamic acid, N-(3-hydroxytricyclo[3.3.1.13,7]dec-1-yl)-, 1,1-dimethylethyl ester (ACI)
IUPAC Name tert-butyl N-(3-hydroxy-1-adamantyl)carbamate
SMILES CC(C)(C)OC(=O)NC12CC3CC(CC(O)(C3)C1)C2
Specifications
Quantity 250 mg
InChI Formula InChI=1S/C15H25NO3/c1-13(2,3)19-12(17)16-14-5-10-4-11(6-14)8-15(18,7-10)9-14/h10-11,18H,4-9H2,1-3H3,(H,16,17)
Formula Weight 267.1834
Chemical Name or Material (3-Hydroxytricyclo[3.3.1.13,7]dec-1-yl)carbamic Acid 1,1-Dimethylethyl Ester