CAS RN 847795-89-5
CAS RN 847795-89-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(3-Hydroxytricyclo[3.3.1.13,7]dec-1-yl)carbamic Acid 1,1-Dimethylethyl Ester, TRC
CAS: 847795-89-5 Molecular Formula: C15H25NO3 Molecular Weight (g/mol): 267.36 Synonym: 1,1-Dimethylethyl N-(3-hydroxytricyclo[3.3.1.13,7]dec-1-yl)carbamate (ACI),Carbamic acid, (3-hydroxytricyclo[3.3.1.13,7]dec-1-yl)-, 1,1-dimethylethyl ester (9CI),Carbamic acid, N-(3-hydroxytricyclo[3.3.1.13,7]dec-1-yl)-, 1,1-dimethylethyl ester (ACI) IUPAC Name: tert-butyl N-(3-hydroxy-1-adamantyl)carbamate SMILES: CC(C)(C)OC(=O)NC12CC3CC(CC(O)(C3)C1)C2