missing translation for 'onlineSavingsMsg'
Learn More

3-Ethoxy-2,4-difluorobenzyl bromide, 97%, Thermo Scientific™

Product Code. 15465187
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
15465187 5 g 5g
15455187 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 15465187

Brand: Thermo Scientific Alfa Aesar H32466.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1017778-22-1
Molecular Formula C9H9BrF2O
Molecular Weight (g/mol) 251.071
MDL Number MFCD09258729
InChI Key BOXYRQBXOYISBL-UHFFFAOYSA-N
Synonym 3-ethoxy-2,4-difluorobenzyl bromide, 1-bromomethyl-3-ethoxy-2,4-difluorobenzene, 3-ethoxy-2,4-difluorobenzyl bromide, jrd, 4-bromomethyl-2-ethoxy-1,3-difluorobenzene
PubChem CID 46737646
IUPAC Name 1-(bromomethyl)-3-ethoxy-2,4-difluorobenzene
SMILES CCOC1=C(C=CC(=C1F)CBr)F

Specifications

Refractive Index 1.5175
Quantity 5 g
UN Number UN3265
Formula Weight 251.07
Percent Purity 97%
Chemical Name or Material 3-Ethoxy-2,4-difluorobenzyl bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.