accessibility menu, dialog, popup

UserName

CAS RN 1017778-22-1

CH3OFBrF

CAS RN 1017778-22-1

IUPAC Name: 1-(bromomethyl)-3-ethoxy-2,4-difluorobenzene
Synonyms: 1-(bromomethyl)-3-ethoxy-2,4-difluorobenzene
Molecular Weight (g/mol): 251.07
Molecular Formula: C9H9BrF2O
InChi Key: BOXYRQBXOYISBL-UHFFFAOYSA-N
SMILES: CCOC1=C(F)C=CC(CBr)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results