Learn More
3-Decen-2-one, 97%, Thermo Scientific™
Product Code. 11429526
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. Quantity unitSize
11429526 5 g 5g
11439526 25 g 25g
2 options
Product Code. 11429526
Brand: Thermo Scientific Alfa Aesar A19256.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 10519-33-2
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.253
MDL Number MFCD00015700
InChI Key JRPDANVNRUIUAB-CMDGGOBGSA-N
Synonym 3-decen-2-one, heptylidene acetone, oenanthylidene acetone, e-3-decen-2-one, unii-z22804bqxd, 3e-dec-3-en-2-one, e-dec-3-en-2-one, dec-3-en-2-one, fema no. 3532, enanthylidene acetone
PubChem CID 5363233
IUPAC Name (E)-dec-3-en-2-one
SMILES CCCCCCC=CC(=O)C
Specifications
Density 0.84
Quantity 5 g
Formula Weight 154.25
Percent Purity 97%
Chemical Name or Material 3-Decen-2-one