Learn More
3-Cyanopyridine 1-oxide, 97%
Product Code. 11418837
Click to view available options
Quantity:
2.5 g
10 g
Unit Size:
10g
2.5g
Product Code. 11418837
Brand: Thermo Scientific Alfa Aesar A19961.88
Request Bulk or Custom Format
CAS: 14906-64-0 | C6H5N2O | 121.12 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 14906-64-0
Molecular Formula C6H5N2O
Molecular Weight (g/mol) 121.12
MDL Number MFCD00129037
InChI Key AABSWFBNBQBPEQ-UHFFFAOYNA-N
Synonym nicotinonitrile-1-oxide, 3-cyanopyridine n-oxide, 3-cyanopyridine-n-oxide, nicotinonitrile 1-oxide, 3-cyanopyridine 1-oxide, 3-cyanopyridin-1-ium-1-olate, 3-pyridinecarbonitrile, 1-oxide, acmc-1c6hr, 3-pyridinecarbonitrile,1-oxide, woovsqcalyyudo-uhfffaoysa
PubChem CID 84686
SMILES [O-][NH++]1C=C[CH-]C(=C1)C#N
Specifications
Melting Point 175°C to 178°C
Quantity 2.5 g
Sensitivity Hygroscopic
IUPAC Name 1-oxidopyridin-1-ium-3-carbonitrile
Formula Weight 120.11
Percent Purity 97%
Chemical Name or Material 3-Cyanopyridine 1-oxide